Assay Details
| Gene Symbol: CASP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015115566.2 | XP_014971052.2 | 1161 | 126 | ||
| XM_028833930.1 | XP_028689763.1 | 1076 | 126 | |||
More Information
Set Membership: |
|

CASP5
Genomic Map