Assay Details
| Gene Symbol: ELOC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198677.1 | NP_001185606.1 | 438 | 193 | ||
| XM_015145513.2 | XP_015000999.1 | 440 | 193 | |||
| XM_015145514.2 | XP_015001000.1 | 460 | 193 | |||
| XM_015145515.2 | XP_015001001.1 | 485 | 193 | |||
| XM_015145516.2 | XP_015001002.1 | 500 | 193 | |||
| XM_028852144.1 | XP_028707977.1 | 608 | 193 | |||
| GenBank mRNA | CN805102.1 | - | 438 | 193 | ||
| JU476975.1 | - | 376 | 193 | |||
| JV048071.1 | - | 381 | 193 | |||
| JV592023.1 | - | 376 | 193 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ELOC
Genomic Map