Assay Details
| Gene Symbol: TUBA1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195391.1 | NP_001182320.1 | 1509 | 141 | ||
| XM_028828777.1 | XP_028684610.1 | 3833 | 141 | |||
| GenBank mRNA | CN646945.1 | - | 753 | 141 | ||
| JV481386.1 | - | 368 | 141 | |||
| JV628316.1 | - | 1512 | 141 | |||
| Gene Symbol: TUBA1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU477146.1 | - | 1531 | 141 | ||
| JV667159.1 | - | 1517 | 141 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TUBA1A
Genomic Map