Assay Details
| Gene Symbol: ANP32B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266908.1 | NP_001253837.1 | 1435 | 144 | ||
| GenBank mRNA | JV045935.1 | - | 1430 | 144 | ||
| JV635699.1 | - | 1435 | 144 | |||
More Information
Set Membership: |
|

ANP32B
Genomic Map