Assay Details
| Gene Symbol: LOC719765 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028835366.1 | XP_028691199.1 | 4037 | 104 | ||
| Gene Symbol: LOC719627 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015117319.2 | XP_014972805.2 | 4041 | 104 | ||
More Information
Set Membership: |
|

LOC719765
Genomic Map