Assay Details
| Gene Symbol: LARP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015142589.2 | XP_014998075.2 | 2442 | 133 | ||
| XM_028851082.1 | XP_028706915.1 | 1842 | 133 | |||
| GenBank mRNA | JU327240.1 | - | 1791 | 133 | ||
| JV512021.1 | - | 1822 | 133 | |||
| JV512022.1 | - | 1817 | 133 | |||
More Information
Set Membership: |
|

LARP6
Genomic Map