Assay Details
| Gene Symbol: TOMM22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU326060.1 | - | 1104 | 95 | ||
| JU328274.1 | - | 1093 | 95 | |||
| JU328275.1 | - | 1099 | 95 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TOMM22| Gene Symbol: TOMM22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU326060.1 | - | 1104 | 95 | ||
| JU328274.1 | - | 1093 | 95 | |||
| JU328275.1 | - | 1099 | 95 | |||
Set Membership: |
|