Assay Details
| Gene Symbol: LOC706568 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015115567.2 | XP_014971053.1 | 990 | 101 | ||
| XM_015115568.2 | XP_014971054.1 | 990 | 101 | |||
| GenBank mRNA | JU325009.1 | - | 327 | 101 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LOC706568
Genomic Map