Assay Details
| Gene Symbol: CD151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001361174.2 | NP_001348103.1 | 166 | 71 | ||
| NM_001361175.2 | NP_001348104.1 | 162 | 71 | |||
| NM_001361176.2 | NP_001348105.1 | 224 | 71 | |||
| NM_001361177.1 | NP_001348106.1 | 229 | 71 | |||
| XM_015113287.2 | XP_014968773.1 | 320 | 71 | |||
| XM_015113289.1 | XP_014968775.1 | 212 | 71 | |||
| XM_028832488.1 | XP_028688321.1 | 311 | 71 | |||
| XM_028832489.1 | XP_028688322.1 | 330 | 71 | |||
| XM_028832490.1 | XP_028688323.1 | 216 | 71 | |||
| XM_028832491.1 | XP_028688324.1 | 183 | 71 | |||
| XM_028832493.1 | XP_028688326.1 | 334 | 71 | |||
| XM_028832494.1 | XP_028688327.1 | 223 | 71 | |||
| GenBank mRNA | AF275665.1 | - | 114 | 71 | ||
| JU471378.1 | - | 164 | 71 | |||
| JV048827.1 | - | 112 | 71 | |||
| JV634758.1 | - | 91 | 71 | |||
| JV638340.1 | - | 176 | 71 | |||
| JV669377.1 | - | 166 | 71 | |||
More Information
Set Membership: |
|

CD151
Genomic Map