Assay Details
| Gene Symbol: ASMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032940.1 | NP_001028112.1 | 734 | 134 | ||
| GenBank mRNA | AY069924.1 | - | 734 | 134 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ASMT| Gene Symbol: ASMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032940.1 | NP_001028112.1 | 734 | 134 | ||
| GenBank mRNA | AY069924.1 | - | 734 | 134 | ||
Set Membership: |
|