Assay Details
| Gene Symbol: CASP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042431.2 | NP_001035896.1 | 284 | 63 | ||
| XM_028832883.1 | XP_028688716.1 | 513 | 63 | |||
| GenBank mRNA | DQ644576.1 | - | 287 | 63 | ||
More Information
Set Membership: |
|

CASP12
Genomic Map