Assay Details
| Gene Symbol: NAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044736.1 | NP_001038201.1 | 543 | 107 | ||
| XM_015144793.2 | XP_015000279.1 | 2499 | 107 | |||
| XM_015144795.2 | XP_015000281.1 | 650 | 107 | |||
More Information
Set Membership: |
|

NAT2
Genomic Map