Assay Details
| Gene Symbol: HBE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047147.1 | NP_001040612.1 | 91 | 95 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HBE1| Gene Symbol: HBE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047147.1 | NP_001040612.1 | 91 | 95 | ||