Assay Details
| Gene Symbol: ADH1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042765.1 | NP_001036230.1 | 280 | 139 | ||
| GenBank mRNA | M81807.1 | - | 280 | 139 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADH1A| Gene Symbol: ADH1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042765.1 | NP_001036230.1 | 280 | 139 | ||
| GenBank mRNA | M81807.1 | - | 280 | 139 | ||
Set Membership: |
|