Assay Details
| Gene Symbol: MNP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032863.3 | NP_001028035.1 | 433 | 134 | ||
| GenBank mRNA | AF184161.1 | - | 327 | 134 | ||
| JU478342.1 | - | 321 | 134 | |||
More Information
Set Membership: |
|

MNP2
Genomic Map