Assay Details
| Gene Symbol: KIR3DL0 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047119.2 | NP_001040584.1 | 352 | 69 | ||
| GenBank mRNA | DQ157756.1 | - | 353 | 69 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIR3DL0| Gene Symbol: KIR3DL0 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047119.2 | NP_001040584.1 | 352 | 69 | ||
| GenBank mRNA | DQ157756.1 | - | 353 | 69 | ||
Set Membership: |
|