Assay Details
| Gene Symbol: KIR2DL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042661.1 | NP_001036126.1 | 237 | 103 | ||
| XM_028838563.1 | XP_028694396.1 | 486 | 103 | |||
| GenBank mRNA | AF334644.1 | - | 181 | 103 | ||
| AF334645.1 | - | 181 | 103 | |||
| AF361088.1 | - | 237 | 103 | |||
| AY505486.1 | - | 181 | 103 | |||
| AY728182.1 | - | 200 | 103 | |||
| EU702486.1 | - | 199 | 103 | |||
| FJ211402.1 | - | 184 | 103 | |||
| FJ211403.1 | - | 184 | 103 | |||
| FJ211404.1 | - | 184 | 103 | |||
| FJ824091.1 | - | 199 | 103 | |||
| FJ824092.1 | - | 199 | 103 | |||
| FJ824093.1 | - | 199 | 103 | |||
| FJ824094.1 | - | 199 | 103 | |||
| FJ824095.1 | - | 199 | 103 | |||
| FJ824096.1 | - | 199 | 103 | |||
| FJ824097.1 | - | 103 | ||||
| FJ824098.1 | - | 199 | 103 | |||
| FJ824099.1 | - | 199 | 103 | |||
| FJ824100.1 | - | 199 | 103 | |||
| FJ824101.1 | - | 199 | 103 | |||
| FJ824102.1 | - | 199 | 103 | |||
| FJ824103.1 | - | 199 | 103 | |||
| FJ824104.1 | - | 199 | 103 | |||
| FJ824105.1 | - | 199 | 103 | |||
| FJ824106.1 | - | 103 | ||||
| FJ824107.1 | - | 199 | 103 | |||
| FJ824108.1 | - | 103 | ||||
| FJ824109.1 | - | 199 | 103 | |||
| FJ824110.1 | - | 199 | 103 | |||
| GU014298.1 | - | 103 | ||||
| GU112263.1 | - | 199 | 103 | |||
| GU112279.1 | - | 199 | 103 | |||
| GU112280.1 | - | 199 | 103 | |||
| GU112284.1 | - | 199 | 103 | |||
| GU112287.1 | - | 199 | 103 | |||
| GU112303.1 | - | 199 | 103 | |||
| GU112313.1 | - | 103 | ||||
| GU112316.1 | - | 199 | 103 | |||
| GU112318.1 | - | 199 | 103 | |||
| GU112322.1 | - | 199 | 103 | |||
| GU112326.1 | - | 199 | 103 | |||
| GU112331.1 | - | 199 | 103 | |||
| GU299490.1 | - | 181 | 103 | |||
| GU564176.1 | - | 178 | 103 | |||
| GU564177.1 | - | 178 | 103 | |||
| GU564178.1 | - | 178 | 103 | |||
| GU564180.1 | - | 178 | 103 | |||
| GU564181.1 | - | 178 | 103 | |||
| KY660350.1 | - | 199 | 103 | |||
| KY660351.1 | - | 199 | 103 | |||
| KY660352.1 | - | 199 | 103 | |||
| KY660353.1 | - | 199 | 103 | |||
| LR030481.1 | - | 199 | 103 | |||
| LR030482.1 | - | 199 | 103 | |||
| LR030483.1 | - | 199 | 103 | |||
| LR030484.1 | - | 199 | 103 | |||
| LR030485.1 | - | 199 | 103 | |||
| LR030486.1 | - | 103 | ||||
| LR030487.1 | - | 199 | 103 | |||
| LR030488.1 | - | 199 | 103 | |||
| LR030489.1 | - | 199 | 103 | |||
| LR030490.1 | - | 199 | 103 | |||
| LR694596.1 | - | 199 | 103 | |||
| LR694597.1 | - | 199 | 103 | |||
| LR694598.1 | - | 199 | 103 | |||
| LR694600.1 | - | 199 | 103 | |||
| LR694601.1 | - | 199 | 103 | |||
| LR694602.1 | - | 199 | 103 | |||
| LR694605.1 | - | 199 | 103 | |||
| LR694606.1 | - | 199 | 103 | |||
| LR694607.1 | - | 199 | 103 | |||
| LR694608.1 | - | 199 | 103 | |||
| LR694609.1 | - | 199 | 103 | |||
| LS997696.1 | - | 199 | 103 | |||
| LS997697.1 | - | 199 | 103 | |||
| LS997698.1 | - | 199 | 103 | |||
| LS997699.1 | - | 103 | ||||
| LS997700.1 | - | 103 | ||||
| LS997701.1 | - | 103 | ||||
| LS997702.1 | - | 103 | ||||
More Information
Set Membership: |
|

KIR2DL4
Genomic Map