Assay Details
| Gene Symbol: WASHC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015152599.2 | XP_015008085.1 | 562 | 113 | ||
| XM_015152600.2 | XP_015008086.1 | 559 | 113 | |||
| XM_015152602.2 | XP_015008088.1 | 591 | 113 | |||
| XM_015152603.2 | XP_015008089.1 | 588 | 113 | |||
| XM_015152604.2 | XP_015008090.1 | 463 | 113 | |||
| GenBank mRNA | JU334760.1 | - | 529 | 113 | ||
| JU473164.1 | - | 522 | 113 | |||
| JV043805.1 | - | 534 | 113 | |||
| JV453124.1 | - | 500 | 113 | |||
More Information
Set Membership: |
|

WASHC3
Genomic Map