Assay Details
| Gene Symbol: NLRP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114114.1 | NP_001107586.1 | 807 | 96 | ||
| XM_028832874.1 | XP_028688707.1 | 915 | 96 | |||
| GenBank mRNA | EU338447.1 | - | 807 | 96 | ||
More Information
Set Membership: |
|

NLRP6
Genomic Map