Assay Details
| Gene Symbol: NLRP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114352.1 | NP_001107824.1 | 329 | 89 | ||
| GenBank mRNA | EU344967.1 | - | 329 | 89 | ||
| JV655647.1 | - | 385 | 89 | |||
More Information
Set Membership: |
|

NLRP2
Genomic Map