Assay Details
| Gene Symbol: APOBEC3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114359.2 | NP_001107831.1 | 458 | 117 | ||
| GenBank mRNA | EU381233.1 | - | 350 | 117 | ||
| JF714486.1 | - | 350 | 117 | |||
| JU334043.1 | - | 438 | 117 | |||
| JU470722.1 | - | 1030 | 117 | |||
| JU475210.1 | - | 447 | 117 | |||
| JV044579.1 | - | 458 | 117 | |||
| JV044581.1 | - | 458 | 117 | |||
| JV058196.1 | - | 327 | 117 | |||
| JV637424.1 | - | 452 | 117 | |||
| JV637426.1 | - | 452 | 117 | |||
More Information
Set Membership: |
|

APOBEC3C
Genomic Map