Assay Details
| Gene Symbol: LOC710848 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001107817.4 | XP_001107817.2 | 467 | 93 | ||
| Gene Symbol: IFNA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194384.1 | NP_001181313.1 | 242 | 93 | ||
| Gene Symbol: IFNA21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001108051.4 | XP_001108051.2 | 93 | |||
| Gene Symbol: LOC106992322 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015129534.2 | XP_014985020.1 | 429 | 93 | ||
| Gene Symbol: LOC114672663 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028834485.1 | XP_028690318.1 | 242 | 93 | ||
| Gene Symbol: LOC114672664 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028834487.1 | XP_028690320.1 | 275 | 93 | ||
| Gene Symbol: LOC710487 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001107576.4 | XP_001107576.2 | 93 | |||
| Gene Symbol: LOC710568 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015117658.2 | XP_014973144.1 | 93 | |||
| Gene Symbol: LOC710751 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015117659.2 | XP_014973145.2 | 93 | |||
| Gene Symbol: LOC711088 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028834486.1 | XP_028690319.1 | 93 | |||
More Information
Set Membership: |
|

LOC710848
Genomic Map