Assay Details
| Gene Symbol: LILRA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040670.2 | NP_001035760.2 | 2747 | 88 | ||
| GenBank mRNA | DQ155427.1 | - | 2747 | 88 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LILRA1| Gene Symbol: LILRA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040670.2 | NP_001035760.2 | 2747 | 88 | ||
| GenBank mRNA | DQ155427.1 | - | 2747 | 88 | ||
Set Membership: |
|