Assay Details
| Gene Symbol: SPIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260981.1 | NP_001247910.1 | 283 | 81 | ||
| GenBank mRNA | JU333214.1 | - | 296 | 81 | ||
| JU474529.1 | - | 283 | 81 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001260981.1 transcripts.
Set Membership: |
|

SPIN3
Genomic Map