Assay Details
Gene Symbol: TRIM77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001096252.2 | XP_001096252.2 | 736 | 86 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
