Assay Details
| Gene Symbol: SOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142940.1 | NP_001136412.1 | 350 | 81 | ||
| XM_028843257.1 | XP_028699090.1 | 3121 | 81 | |||
| XM_028843258.1 | XP_028699091.1 | 3618 | 81 | |||
| XM_028843259.1 | XP_028699092.1 | 2870 | 81 | |||
| GenBank mRNA | FJ375750.1 | - | 350 | 81 | ||
| JU336054.1 | - | 416 | 81 | |||
More Information
Set Membership: |
|

SOX2
Genomic Map