Assay Details
| Gene Symbol: SELENOP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159490.2 | NP_001152962.1 | 675 | 94 | ||
| GenBank mRNA | JV601665.1 | - | 483 | 94 | ||
| JV693087.1 | - | 495 | 94 | |||
| JV721001.1 | - | 416 | 94 | |||
More Information
Set Membership: |
|

SELENOP
Genomic Map