Assay Details
| Gene Symbol: SPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125936.2 | XP_014981422.2 | 1122 | 90 | ||
| GenBank mRNA | JU492142.1 | - | 317 | 90 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPN| Gene Symbol: SPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125936.2 | XP_014981422.2 | 1122 | 90 | ||
| GenBank mRNA | JU492142.1 | - | 317 | 90 | ||
Set Membership: |
|