Assay Details
| Gene Symbol: LOC114669827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028826636.1 | XP_028682469.1 | 467 | 98 | ||
| GenBank mRNA | JU334336.1 | - | 98 | |||
| JV715441.1 | - | 374 | 98 | |||
| Gene Symbol: IFIT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015147649.2 | XP_015003135.2 | 98 | |||
| XM_028826634.1 | XP_028682467.1 | 98 | ||||
| XM_028826635.1 | XP_028682468.1 | 98 | ||||
| GenBank mRNA | JU334337.1 | - | 98 | |||
| JV046955.1 | - | 98 | ||||
| Gene Symbol: LOC722130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU325264.1 | - | 175 | 98 | ||
More Information
Set Membership: |
|

LOC114669827
Genomic Map