Assay Details
| Gene Symbol: KCNS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168626.2 | NP_001162097.1 | 478 | 87 | ||
| XM_015148832.2 | XP_015004318.2 | 586 | 87 | |||
| GenBank mRNA | JV565114.1 | - | 84 | 87 | ||
| JV581208.1 | - | 84 | 87 | |||
More Information
Set Membership: |
|

KCNS1
Genomic Map