Assay Details
| Gene Symbol: JDP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015144150.2 | XP_014999636.1 | 377 | 83 | ||
| XM_015144151.2 | XP_014999637.1 | 530 | 83 | |||
| XM_028851699.1 | XP_028707532.1 | 243 | 83 | |||
| XM_028851700.1 | XP_028707533.1 | 465 | 83 | |||
| GenBank mRNA | JU338615.1 | - | 262 | 83 | ||
| JV672611.1 | - | 261 | 83 | |||
More Information
Set Membership: |
|

JDP2
Genomic Map