Assay Details
| Gene Symbol: IER5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002802002.3 | XP_002802048.3 | 920 | 80 | ||
| GenBank mRNA | JV618660.1 | - | 713 | 80 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

IER5| Gene Symbol: IER5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002802002.3 | XP_002802048.3 | 920 | 80 | ||
| GenBank mRNA | JV618660.1 | - | 713 | 80 | ||