Assay Details
| Gene Symbol: IZUMO1R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193805.1 | NP_001180734.1 | 330 | 89 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

IZUMO1R| Gene Symbol: IZUMO1R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193805.1 | NP_001180734.1 | 330 | 89 | ||