Assay Details
| Gene Symbol: SOCS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193820.2 | NP_001180749.1 | 2444 | 89 | ||
| XM_001087968.4 | XP_001087968.1 | 2666 | 89 | |||
| XM_015143704.2 | XP_014999190.1 | 2788 | 89 | |||
| XM_015143705.2 | XP_014999191.1 | 2784 | 89 | |||
| GenBank mRNA | JU473023.1 | - | 2707 | 89 | ||
| JU473024.1 | - | 2703 | 89 | |||
More Information
Set Membership: |
|

SOCS4
Genomic Map