Assay Details
| Gene Symbol: INSYN2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193840.3 | NP_001180769.2 | 289 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001193840.3 transcripts.
Set Membership: |
|

INSYN2A
Genomic Map