Assay Details
| Gene Symbol: INSYN2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193840.3 | NP_001180769.2 | 1811 | 90 | ||
| XM_015148446.2 | XP_015003932.2 | 1447 | 90 | |||
| XM_028853305.1 | XP_028709138.1 | 1364 | 90 | |||
| XM_028853306.1 | XP_028709139.1 | 1290 | 90 | |||
| GenBank mRNA | JV721088.1 | - | 249 | 90 | ||
More Information
Set Membership: |
|

INSYN2A
Genomic Map