Assay Details
| Gene Symbol: INSYN2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193931.2 | NP_001180860.2 | 466 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001193931.2 transcripts.
Set Membership: |
|

INSYN2B
Genomic Map