Assay Details
| Gene Symbol: INSYN2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193931.2 | NP_001180860.2 | 2812 | 102 | ||
| Gene Symbol: LOC114675390 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028842651.1 | XP_028698484.1 | 2881 | 102 | ||
More Information
Set Membership: |
|

INSYN2B
Genomic Map