Assay Details
| Gene Symbol: EDDM3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193940.2 | NP_001180869.1 | 79 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001193940.2 transcripts.
Set Membership: |
|

EDDM3B
Genomic Map