Assay Details
| Gene Symbol: FABP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193941.2 | NP_001180870.2 | 80 | 82 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FABP9| Gene Symbol: FABP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193941.2 | NP_001180870.2 | 80 | 82 | ||