Assay Details
| Gene Symbol: KRT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193952.2 | NP_001180881.2 | 749 | 73 | ||
| GenBank mRNA | JU472287.1 | - | 827 | 73 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT7| Gene Symbol: KRT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193952.2 | NP_001180881.2 | 749 | 73 | ||
| GenBank mRNA | JU472287.1 | - | 827 | 73 | ||
Set Membership: |
|