Assay Details
| Gene Symbol: DPEP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194056.1 | NP_001180985.1 | 1213 | 100 | ||
| GenBank mRNA | JV068471.1 | - | 1167 | 100 | ||
| JV635684.1 | - | 1227 | 100 | |||
More Information
Set Membership: |
|

DPEP3
Genomic Map