Assay Details
| Gene Symbol: DPEP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194056.1 | NP_001180985.1 | 1349 | 90 | ||
| GenBank mRNA | JV068471.1 | - | 1303 | 90 | ||
| JV635684.1 | - | 1363 | 90 | |||
More Information
Set Membership: |
|

DPEP3
Genomic Map