Assay Details
| Gene Symbol: FAM127A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194127.1 | NP_001181056.1 | 52 | 78 | ||
| GenBank mRNA | JU336960.1 | - | 49 | 78 | ||
| JU336962.1 | - | 49 | 78 | |||
| JU336963.1 | - | 49 | 78 | |||
| JU336964.1 | - | 49 | 78 | |||
| JU336965.1 | - | 49 | 78 | |||
| JU336966.1 | - | 49 | 78 | |||
| JU477695.1 | - | 49 | 78 | |||
| JU477696.1 | - | 49 | 78 | |||
| JU477697.1 | - | 49 | 78 | |||
| JV632528.1 | - | 52 | 78 | |||
| JV632530.1 | - | 52 | 78 | |||
| JV632535.1 | - | 52 | 78 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194127.1 transcripts.
Set Membership: |
|

FAM127A
Genomic Map