Assay Details
| Gene Symbol: EID2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194175.1 | NP_001181104.1 | 53 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194175.1 transcripts.
Set Membership: |
|

EID2
Genomic Map