Assay Details
| Gene Symbol: EID2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194180.2 | NP_001181109.2 | 753 | 104 | ||
| GenBank mRNA | JV642099.1 | - | 735 | 104 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EID2B| Gene Symbol: EID2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194180.2 | NP_001181109.2 | 753 | 104 | ||
| GenBank mRNA | JV642099.1 | - | 735 | 104 | ||
Set Membership: |
|