Assay Details
| Gene Symbol: HSCB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194228.1 | NP_001181157.1 | 378 | 74 | ||
| XM_028827360.1 | XP_028683193.1 | 413 | 74 | |||
| XM_028827361.1 | XP_028683194.1 | 409 | 74 | |||
| XM_028827362.1 | XP_028683195.1 | 409 | 74 | |||
| XM_028827363.1 | XP_028683196.1 | 408 | 74 | |||
| XM_028827364.1 | XP_028683197.1 | 351 | 74 | |||
| XM_028827365.1 | XP_028683198.1 | 93 | 74 | |||
| GenBank mRNA | JU325710.1 | - | 313 | 74 | ||
| JV046269.1 | - | 342 | 74 | |||
| JV572349.1 | - | 344 | 74 | |||
| JV635541.1 | - | 333 | 74 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028827364.1, XM_028827365.1 transcripts.
Set Membership: |
|

HSCB
Genomic Map