Assay Details
| Gene Symbol: C20H16orf72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194229.2 | NP_001181158.1 | 294 | 80 | ||
| GenBank mRNA | JU319631.1 | - | 247 | 80 | ||
| JU475279.1 | - | 440 | 80 | |||
| JV504087.1 | - | 248 | 80 | |||
| JV579513.1 | - | 271 | 80 | |||
| JV601007.1 | - | 550 | 80 | |||
More Information
Set Membership: |
|

C20H16orf72
Genomic Map