Assay Details
| Gene Symbol: TENT5D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194232.1 | NP_001181161.1 | 121 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194232.1 transcripts.
Set Membership: |
|

TENT5D
Genomic Map