Assay Details
| Gene Symbol: P2RY12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194370.2 | NP_001181299.1 | 293 | 73 | ||
| XM_015131755.2 | XP_014987241.1 | 317 | 73 | |||
| GenBank mRNA | JU322372.1 | - | 174 | 73 | ||
| JV721319.1 | - | 212 | 73 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194370.2, XM_015131755.2 transcripts.
Set Membership: |
|

P2RY12
Genomic Map